Learn More
Tocris Bioscience™ PD 4'-piperazine
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
PD 4'-piperazine is a cereblon ligand that can be used for PROTAC® Degrader development. The compound can be used to develop PD-PROTAC molecules that exhibit improved stability and potency compared with PROTACs synthesized using IMiD (immunomodulatory imide drug) analogs.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | Dihydro-1-[4-(1-piperazinyl)phenyl]-2,4(1H,3H)-pyrimidinedione dihydrochloride |
| Quantity | 50 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C14H18N4O2.2HCl |