Learn More
Abnova™ PCSK4 (Human) Recombinant Protein
Human PCSK4 full-length ORF ( AAH36354, 1 a.a. - 242 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | AAH36354 |
---|---|
Gene ID (Entrez) | 54760 |
Name | proprotein convertase subtilisin/kexin type 4 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16160733
|
Abnova™
H00054760-P01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16170733
|
Abnova™
H00054760-P01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Description
- Sequence: MGTRSTLVAIRPLDVSTEGYNNWVFMSTHFWDENPQGVWTLGLENKGYYFNTGTLYRYTLLLYGTAEDMTARPTGPQVTSSACVQRDTEGLCQACDGPAYILGQLCLAYCPPRFFNHTRLVTAGPGHTAAPALRVCSSCHASCYTCRGGSPRDCTSCPPSSTLDQQQGSCMGPTTPDSRPRLRAAACPHHRCPASAMVLSLLAVTLGGPVLCGMSMDLPLYAWLSRARATPTKPQVWLPAGT
Specifications
AAH36354 | |
proprotein convertase subtilisin/kexin type 4 | |
125% SDS-PAGE Stained with Coomassie Blue | |
DKFZp434B217, MGC34749, PC4, SPC5 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
54760 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PCSK4 | |
GST |