Learn More
BRAND™ 12-Strip PCR Tubes, BIO-CERT™ PCR Quality
Product Code. 10673113 Shop All Brand Products
Click to view available options
Color:
Clear
Yellow
Unit Size:
Pack of 125
Pack of 80
Product Code. 10673113
Brand: BRAND™ 781280
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Manufactured from high-quality virgin plastics for excellent, reproducible results. Domed caps available separately.
BRAND™ PCR Tube Strips offer exceptional and versatile performance for PCR or qPCR setups, medium-throughput applications, and other workflows requiring precise thermal transfer and high cleanliness. Made from high-quality virgin polypropylene, these tubes ensure reliable results with features that enhance usability and sample integrity. Key features include:
  • BIO-CERT™ PCR QUALITY: Certified free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors, ensuring the highest level of cleanliness for sensitive PCR applications.
  • Superior Heat Transfer: Extra thin-walled tubes optimize thermal transfer during PCR, reducing cycle times and providing consistent performance over a wide temperature range.
  • User-Friendly Features: Numbered wells provide clear and easy identification.
  • Individually Attached Flat Caps: Feature three connectors for added rigidity, making them ideal for real-time qPCR.
  • Individually Attached Domed Caps: Equipped with a single connector, allowing for easy separation and convenient use.
  • High-Quality Construction: Manufactured from high-quality virgin resins without lubricants or chemical additives.
  • Autoclavable: Designed to withstand autoclaving at 121°C (2 bar) in compliance with DIN EN 285.
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Breakapart No
Binding Property None
Certifications/Compliance Certified RNase/DNase-free
Certified DNA, Dnase-, RNase, and Pyrogen free
Format Strip Plate
Array 1 x 12
Autoclavable Autoclavable
Color Clear
Disposable Single-use
Product Type 12-Tube Strips
For Use With (Equipment) Heat blocks, thermal cyclers
Material Polypropylene
Volume (Metric) Well 0.2 mL
No. of Wells 12
Volume (Metric) Working 0.2 mL
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 0.2 mL
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) PCR, qPCR, thermal cycling
Includes 80 strips of 12 tubes
Quantity 80 Strips
RNase Free Yes