Learn More
DiscoverX pCMV-ARMS1-ProLinkTM 2 Vector
Product Code. 17393269
Encompass
Click to view available options
Quantity:
1 Vector
Unit Size:
1 set
Product Code. 17393269
Brand: DiscoverX 930489
The pCMV-ARMS1-ProLinkTM 2 Vector is a mammalian cloning vector to produce cells expressing a PK2-tagged protein of interest. Contains phosphorylation sites to increase arrestin recruitment. Provided as a vial of 10μg vector in liquid buffer.
The pCMV-ARMS1-ProLinkTM 2 Vector is a mammalian cloning vector for use in expressing a protein of interest fused to the ProLink 2 (PK 2) tag at the C-terminus. The PK tag is a version of ED used for EFC assays (in combination with EA). The ProLink 2 tag has a higher affinity for EA, making it ideal for use with weaker protein:protein interactions.The ARMS (Arrestin Recruitment Modifying Sequence) PK tags contain GRK phosphorylation sites to improve ligand-dependent arrestin recruitment to GPCRs and improve assay performance. Provided as a vial of 10μg vector in 20μL liquid buffer.
Specifications
Product Type Cloning Vector
Quantity 1 Vector
For Use With (Application) Cloning Vectors
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.