Learn More
Emma Safety Footwear Patrick XD Safety Shoes
Product Code. 15998961
Change view
Click to view available options
Size:
37
38
39
40
41
42
45
46
48
Unit Size:
1 pair
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998961 48 1 pair
15918961 37 1 pair
15928961 38 1 pair
15938961 39 1 pair
15948961 40 1 pair
15958961 41 1 pair
15968961 42 1 pair
15978961 45 1 pair
15988961 46 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15998961 Supplier Emma Safety Footwear Supplier No. MM73186648
  • ONLINE EXCLUSIVE PRICE
    154.32€
    90.30€ / 1 pairs
    Save 64.02€
    41% Off
Estimated Shipment: 25-03-2027
An ideal choice when optimal foot protection is required. A strong, solid steel plate in the sole and resilient leather uppers defend the feet from unexpected hazards. Perfectly balanced, anatomical arch support will keep feet feeling fit and fresh.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Design Patrick XD
Material Leather
Size 48