Learn More
Intelsius™ PathoShield™
Product Code. 17378599
Encompass_Preferred
Click to view available options
Size:
4
5
8
Dimensions (LxWxH):
191 x 145 x 22 mm
275 x 179 x 43 mm
291 x 191 x 102 mm
Unit Size:
Pack of 25
Pack of 50
Product Code. 17378599
Brand: Intelsius™ PA028
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The PathoShield™ is a complete shipping solution designed for the transportation of Category A and Category B biological materials. Each system is pre-printed with the required markings and includes all the components required to transport your sample safely.
  • Complete Category B shipping solution includes flexible 95kPa tested compliant secondary, absorbent, labels, and relevant UN markings
  • Range of sizes suitable for shipping through postal and courier networks
  • Suitable for a wide range of primaries including blood vials, swabs and sample containers
  • Supplied flat packed for efficient storage and shipping prior to use
Specifications
Length (English) Exterior 7.5 in.
Width (English) Exterior 5.7 in.
Includes Pouch
Size 8
Height (English) Exterior 0.9 in.
Type Sample Transport Packaging
Dimensions (L x W) Pouch: 150 x 190 mm (5.9 x 7.4 in.)
Dimensions (LxWxH) 191 x 145 x 22 mm
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.