Learn More
DiscoverX PathHunter™ U2OS TORC2 Nuclear Translocation Cell Line
Description
The PathHunter™ U2OS TORC2 Nuclear Translocation Cell Line is a stable, engineered cell line for use in studying drug candidates affecting the CRTC/CREB pathway. This cell line enables assessment of ligand (e.g. forskolin) based pathway activation via detection of resulting TORC2 (CRTC2) nuclear translocation. This product uses EFC technology. The included cell line overexpresses PL-tagged TORC2 (CRTC2) and nuclear localized EA. Upon pathway activation, induced nuclear translocation of TORC2 leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | TORC2 (CRTC2) nuclear translocation |
| Cell Line | U2OS |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Forskolin |
| Quantity | 2 Vials |
| Target Entry | TORC2 (CRTC2) |
| Accession Number | NM_181715 |