missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS TGFBR1/TGFBR2/ENG Dimerization Cell Line
Description
The PathHunter™ TGFBR1/TGFBR2/ENG Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. TGF-β1) induced dimerization of TGFBR1/TGFBR2/ENG receptors. This product uses EFC technology. The cell line included overexpresses TGFBR1 target protein fused to PK alongside TGFBR2 target protein fused to EA and untagged ENG. Upon ligand-induced dimerization of TGFBR1/TGFBR2/ENG , detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Dimerization |
| Cell Line | U2OS |
| For Use With (Application) | Cell Lines |
| Primary Agonist | TGF-β1 |
| Quantity | 2 Vials |
| Target Entry | TGFBR1/TGFBR2/ENG |
| Accession Number | NM_004612 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?