missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS RANK-IκB Functional Assay
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The PathHunter™ U2OS RANK-IκB Functional Assay is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting the NFκB pathway. This cell line enables detection of IκB levels, allowing for assessment of NFκB pathway activation-induced IκB degradation. This product uses EFC technology. The cell line included overexpresses IκB target protein fused to PL alongside untagged RANK. EA is contained in the detection reagent. Upon pathway-activation-induced degradation of IκB EFC signal is lost. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | NFκB pathway activation-induced IκB degradation |
| Cell Line | U2OS |
| Format | 2 vials |
| For Use With (Application) | Cell Lines |
| Primary Agonist | sRANKL |
| Quantity | 2 Vials |
| Target Entry | RANK-IκB |
| Accession Number | NM_003839.3 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction