Learn More
DiscoverX PathHunter™ U2OS mPD-L1 Ligand Cell Line
Product Code. 17354429
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Product Code. 17354429
Brand: DiscoverX 931159C3
The PathHunter™ U2OS mPD-L1 Ligand Cell Line works with Checkpoint signaling cell lines to induce SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells.
The PathHunter U2OS mPD-L1 Ligand cell line is a stable, engineered cell line designed for use with mPD-L1 Checkpoint Signaling Cell Lines. When co-cultured, the mPD-L1 expressed in this cell line will induce activity of mPD-L1 receptors, which may be assayed through SH2 domain protein recruitment. This product is a companion product designed to work with Immunoglobulin Superfamily Checkpoint Assays. The cell line included overexpresses mPD-L1 cell surface ligand. When co-cultured with the corresponding Checkpoint Signaling Cell Line, the mPD-L1 ligand binds to the checkpoint receptor, inducing receptor activity. This receptor activity leads to recruitment of the relevant SH2 domain protein to the checkpoint receptor inside the receptor cell line and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Assay Entry Ligand Presenting
Cell Line U2OS
For Use With (Application) Cell Lines
Primary Agonist Anti-mPD-L1 Antibody
Quantity 2 Vials
Accession Number NM_021893
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.