missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ U2OS mFCGR2B Clustering Cell Line

Product Code. 17304419
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
This item is not returnable. View return policy

Product Code. 17304419

Brand: DiscoverX 931139C3

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ U2OS mFCGR2B Clustering Cell Line is used to determine cross-linking based potency increases of agonist antibodies. This cell line ships as two vials of cryopreserved cells. Mouse version.

The Path Hunter mFcyRIIb clustering cell line is a stable, engineered cell line designed to increase signal produced from a Checkpoint Cell Line via antibody-mediated crosslinking of the receptors. This product is a companion product used to determine cross-linking based increases in potency of agonist antibodies. The cell line included overexpresses mFcyRIIb Fcg receptor. When co-cultured with a Checkpoint Cell Line in the presence of agonist antibody, the Fcg receptor binds to the antibody, inducing receptor clustering. The induced receptor clustering leads to increased activation and downstreamsignaling, leading to increased EFC signal production. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Mouse version of this protein.

Specifications

Assay Entry Receptor Clustering
Cell Line U2OS
For Use With (Application) Cell Lines
Quantity 2 Vials
Target Entry mFcyRIIb
Accession Number NM_001077189.1
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.