missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ U2OS LCK Activity Assay

Product Code. 17374379
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17374379 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17374379 Supplier DiscoverX Supplier No. 931100C3

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ U2OS LCK Activity Assay measures LCK CTK activity via ligand-induced SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells.

The PathHunter™ U2OS LCK Activity Assay is a stable, engineered cell line for use in studying small molecule drug candidates targeting CTKs. This cell line enables assessment of CTK activation via detection of resulting GHR phosphorylation-induced SH2 domain protein recruitment, mediated by LCK-JAK2 fusion protein. This product uses EFC technology. The included cell line overexpresses PK-tagged relevant cytokine receptor and EA-tagged SH2 domain protein alongside an engineered fusion of the cytokine receptor binding domain of JAK2 (Janus Kinase 2) to the catalytic domain from the CTK of interest, LCK. Upon activation of the cytokine receptor, the CRBD-CTK fusion phosphorylates the receptor, leading to SH2-EA recruitment and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry SH2 Recruitment
Cell Line U2OS
For Use With (Application) Cell Lines
Primary Agonist Recombinant Human Growth Hormone
Quantity 2 Vials
Target Entry LCK
Accession Number NM_000163
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.