missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ U2OS IL1RL1/IL1RAP Dimerization Cell Line

Product Code. 17394329
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17394329 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17394329 Supplier DiscoverX Supplier No. 931067C3

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ U2OS IL1RL1/IL1RAP Dimerization Cell Line measures ligand-induced dimerization of IL1RL1/IL1RAP . This cell line ships as two vials of cryopreserved cells.

The PathHunter™ IL1RL1/IL1RAP Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-33) induced dimerization of IL1RL1/IL1RAP receptors. This product uses EFC technology. The cell line included overexpresses IL1RL1 target protein fused to PK alongside IL1RAP target protein fused to EA. Upon ligand-induced dimerization of IL1RL1/IL1RAP, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Dimerization
Cell Line U2OS
For Use With (Application) Cell Lines
Primary Agonist IL-33
Quantity 2 Vials
Target Entry IL1RL1/IL1RAP
Accession Number NM_016232.4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.