missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS FGFR4-β-Klotho Functional Assay
Description
The PathHunter™ U2OS FGFR4-β-Klotho Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated FGFR4 and β-Klotho dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged FGFR4 and an EA-tagged SH2 domain protein. It also overexpresses an untagged coreceptor, β-Klotho Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Receptor Activation |
| Cell Line | U2OS |
| For Use With (Application) | Cell Lines |
| Primary Agonist | FGF-19 |
| Quantity | 2 Vials |
| Target Entry | FGFR4-β-Klotho |
| Accession Number | NM_002011 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?