missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ U2OS FcγRIIa Clustering Cell Line

Product Code. 17314419
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17314419 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17314419 Supplier DiscoverX Supplier No. 931140C3

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ U2OS FcγRIIa Clustering Cell Line is used to determine cross-linking based potency increases of agonist antibodies. This cell line ships as two vials of cryopreserved cells.

The Path Hunter FcγRIIa clustering cell line is a stable, engineered cell line designed to increase signal produced from a Checkpoint Cell Line via antibody-mediated crosslinking of the receptors. This product is a companion product used to determine cross-linking based increases in potency of agonist antibodies. The cell line included overexpresses FcγRIIa Fcg receptor. When co-cultured with a Checkpoint Cell Line in the presence of agonist antibody, the Fcg receptor binds to the antibody, inducing receptor clustering. The induced receptor clustering leads to increased activation and downstreamsignaling, leading to increased EFC signal production. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Receptor Clustering
Cell Line U2OS
For Use With (Application) Cell Lines
Quantity 2 Vials
Target Entry FcγRIIa
Accession Number NM_001136219.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.