missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS CSF1R/CSF1R Dimerization Cell Line
Description
The PathHunter™ CSF1R/CSF1R Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. M-CSF) induced dimerization of CSF1R/CSF1R receptors. This product uses EFC technology. The cell line included overexpresses CSF1R target protein fused to PK alongside CSF1R target protein fused to EA. Upon ligand-induced dimerization of CSF1R/CSF1R , detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Dimerization |
| Cell Line | U2OS |
| For Use With (Application) | Cell Lines |
| Primary Agonist | M-CSF |
| Quantity | 2 Vials |
| Target Entry | CSF1R/CSF1R |
| Accession Number | NM_005211.3 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?