missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ U2OS cINSRa Functional Assay

Product Code. 17314239
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
This item is not returnable. View return policy

Product Code. 17314239

Brand: DiscoverX 931016C3

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ U2OS cINSRa Functional Assay measures cINSRa receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells. Cyno version.

The PathHunter™ U2OS cINSRa Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated cINSRa dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged cINSRa and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the cyno version of the protein.

Specifications

Assay Entry SH2 Recruitment
Cell Line U2OS
For Use With (Application) Cell Lines
Primary Agonist Insulin, Human
Quantity 2 Vials
Target Entry cINSRa
Accession Number XM_005587742.1
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.