missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ U2OS CDC25A Degradation Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The PathHunter™ U2OS CDC25A Degradation Cell Line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting the cell cycle checkpoint pathway. This cell line enables detection of levels of CDC25a, allowing for assessment of CHK-1-activation-induced CDC25a degradation. This product uses EFC technology. The cell line included overexpresses CDC25a target protein fused to PL. EA is contained in the detection reagent. Upon pathway-activation-induced degradation of CDC25a EFC signal is lost. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | CDC25A Degradation |
| Cell Line | U2OS |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Doxorubicin Hydrochloride |
| Quantity | 2 Vials |
| Target Entry | CDC25A |
| Accession Number | NM_001789.2 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction