missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ NCI-H460 CDC25A Degradation Cell Line
Description
The PathHunter™ NCI-H460 CDC25A Degradation Cell Line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting the cell cycle checkpoint pathway. This cell line enables detection of levels of CDC25a, allowing for assessment of CHK-1-activation-induced CDC25a degradation. This product uses EFC technology. The cell line included overexpresses CDC25a target protein fused to PL. EA is contained in the detection reagent. Upon pathway-activation-induced degradation of CDC25a EFC signal is lost. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | CDC25A Degradation |
| Cell Line | NCI-H460 |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Doxorubicin Hydrochloride |
| Quantity | 2 Vials |
| Target Entry | CDC25A |
| Accession Number | NM_001789.2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?