missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ Jurkat CD47-Presenting Cell Line

Product Code. 17394419
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17394419 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17394419 Supplier DiscoverX Supplier No. 931149C19
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The PathHunter™ Jurkat CD47-Presenting Cell Line works with Checkpoint signaling cell lines to induce SH2 domain protein recruitment. This cell line ships as two vials of cryopreserved cells.

The PathHunter CD47 Ligand cell line is a stable, engineered cell line designed for use with SIRPα Checkpoint Signaling Cell Lines. When co-cultured, the CD47 expressed in this cell line will induce activity of SIRPα receptors, which may be assayed through SH2 domain protein recruitment. This product is a companion product designed to work with Immunoglobulin Superfamily Checkpoint Assays. The cell line included overexpresses CD47 cell surface ligand. When co-cultured with the corresponding Checkpoint Signaling Cell Line, the CD47 ligand binds to the checkpoint receptor, inducing receptor activity. This receptor activity leads to recruitment of the relevant SH2 domain protein to the checkpoint receptor inside the receptor cell line and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Ligand Presenting
Cell Line Jurkat
For Use With (Application) Cell Lines
Quantity 2 Vials
Target Entry CD47
Accession Number NM_001777.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.