missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ Insulin Lispro Bioassay Kit

Product Code. 17393219
Encompass
Change view
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17393219 200 dp (2 x 96-well) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17393219 Supplier DiscoverX Supplier No. 930466Y300009

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ Insulin Lispro Bioassay Kit measures INSRb signal by recruitment of SH2 domain protein to the activated receptor. Bioassay kits contain all assay materials: cells, reagents, and plates.

The PathHunter™ Insulin Lispro Bioassay kit provides an easy-to-use cell based assay to determine Insulin Lispro potency and detect neutralizing antibodies. This bioassay assesses insulin or insulin lispro based activation of INSRb RTK receptors via detection of SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged INSRb and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.

Specifications

Assay Entry SH2 Recruitment
For Use With (Application) Assay-Ready Bioassay Kits
Quantity 200 dp (2 x 96-well)
Target Entry INSRb
Cell Line U2OS
Target INSRb
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.