missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ GPR35 Activated GPCR Internalization U2OS Cell Line
Description
The PathHunter™ GPR35 Activated GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line enables assessment of ligand (e.g. LPA, zaprinast) based internalization of GPR35 via detection of localization to the early endosome. This product uses EFC technology. The included cell line overexpresses EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged GPR35. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting GPR35-arrestin complex is internalized to the early endosome, leading to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Receptor Internalization |
| Cell Line | U2OS |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Zaprinast |
| Quantity | 2 Vials |
| Target Entry | GPR35 |
| Accession Number | NM_005301 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?