missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress rCHRM4 CHO-K1 β-Arrestin GPCR Assay
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ rCHRM4 β-Arrestin eXpress Kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand (e.g. acetylcholine) based activation of rCHRM4 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged rCHRM4 and EA-tagged β-Arrestin 2. Activation of rCHRM4 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Rat version of this protein.
Specifications
Specifications
| Assay Entry | β-Arrestin-2 recruitment |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | Acetylcholine |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | rCHRM4 |
| Cell Line | CHO-K1 |
| Target | rCHRM4 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction