missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ EDG5 Activated GPCR Internalization U2OS Cell Line

Product Code. 17393729
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
This item is not returnable. View return policy

Product Code. 17393729

Brand: DiscoverX 930735C3

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ EDG5 Activated GPCR Internalization U2OS Cell Line measures EDG5 (GPCR) internalization via early-endosome localization. This cell line ships as two vials of cryopreserved cells.

The PathHunter™ EDG5 Activated GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line enables assessment of ligand (e.g. S1P) based internalization of EDG5 via detection of localization to the early endosome. This product uses EFC technology. The included cell line overexpresses EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged EDG5. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting EDG5-arrestin complex is internalized to the early endosome, leading to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry Receptor Internalization
Cell Line U2OS
For Use With (Application) Cell Lines
Primary Agonist Sphingosine 1-phosphate
Quantity 2 Vials
Target Entry EDG5
Accession Number NM_004230.3
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.