Learn More
DiscoverX PathHunter™ DLD1 IL7R/CRLF2 Dimerization Cell Line
Product Code. 17304249
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Product Code. 17304249
Brand: DiscoverX 931019C13
The PathHunter™ DLD1 IL7R/CRLF2 Dimerization Cell Line measures ligand-induced dimerization of IL7R/CRLF2 . This cell line ships as two vials of cryopreserved cells.
The PathHunter™ IL7R/CRLF2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. TSLP) induced dimerization of IL7R/CRLF2 receptors. This product uses EFC technology. The cell line included overexpresses IL7R target protein fused to PK alongside CRLF2 target protein fused to EA. Upon ligand-induced dimerization of IL7R/CRLF2, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Assay Entry Dimerization
Cell Line DLD1
For Use With (Application) Cell Lines
Primary Agonist TSLP
Quantity 2 Vials
Target Entry IL7R/CRLF2
Accession Number NM_002185.3
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.