Learn More
DiscoverX PathHunter™ CHO-K1 TAAR5 β-Arrestin Orphan GPCR Cell Line
Product Code. 17313709
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Product Code. 17313709
Brand: DiscoverX 930718C2
The PathHunter™ CHO-K1 TAAR5 β-Arrestin Orphan GPCR Cell Line measures TAAR5 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.
The PathHunter™ TAAR5 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. methylamines) based activation of TAAR5 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged TAAR5 and EA-tagged β-Arrestin 2. Activation of TAAR5 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Assay Entry β-Arrestin recruitment
Cell Line CHO-K1
For Use With (Application) Cell Lines
Quantity 2 Vials
Target Entry TAAR5
Accession Number NM_003967.2
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.