missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ CHO-K1 RARα Protein Interaction Cell Line
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
Description
The PathHunter™ RARα Protein Interaction cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting NHR activation and recruitment of cofactors. This cell line enables assessment of ligand (e.g. retinoic acid) based NHR activation via detection of SRCP (steroid receptor co-activator peptide) recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged RARα NHR and EA-tagged steroid receptor co-activator peptide (SRCP). Upon activation of RARα, induced interaction with SRCP leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Protein Interaction |
| Cell Line | CHO-K1 |
| For Use With (Application) | Cell Lines |
| Primary Agonist | 9-cis-Retinoic acid |
| Quantity | 2 Vials |
| Target Entry | RARα |
| Accession Number | NM_000964 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction