missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ CHO-K1 MR Protein Interaction Cell Line
Description
The PathHunter™ MR Protein Interaction cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting NHR activation and recruitment of cofactors. This cell line enables assessment of ligand (e.g. aldosterone) based NHR activation via detection of SRCP (steroid receptor co-activator peptide) recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged MR NHR and EA-tagged steroid receptor co-activator peptide (SRCP). Upon activation of MR, induced interaction with SRCP leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Specifications
Specifications
| Assay Entry | Receptor Activation |
| Cell Line | CHO-K1 |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Aldosterone |
| Quantity | 2 Vials |
| Target Entry | MR |
| Accession Number | NM_000901 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?