missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ CHO-K1 mPTGIR β-Arrestin Cell Pool

Product Code. 17303539
Encompass
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
This item is not returnable. View return policy

Product Code. 17303539

Brand: DiscoverX 930615P2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ CHO-K1 mPTGIR β-Arrestin Cell Pool measures mPTGIR (GPCR) activity via recruitment of β-Arrestin2. This cell pool ships as two vials of cryopreserved cells. Mouse protein.

The PathHunter™ mPTGIR cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin I2, beraprost) based activation of mPTGIR GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged mPTGIR and EA-tagged β-Arrestin 2. Activation of mPTGIR stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable cell pool is provided as two vials of cryopreserved cells. Please note that this product expresses the mouse version of this protein.

Specifications

Assay Entry β-Arrestin recruitment
Cell Line CHO-K1
For Use With (Application) Cell Pools
Quantity 2 Vials
Target Entry mPTGIR
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.