missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ CHO-K1 GPR34 β-Arrestin Cell Line

Product Code. 17364419
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
2 ampoules
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17364419 2 Vials 2 ampoules
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17364419 Supplier DiscoverX Supplier No. 931146C2
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The PathHunter™ CHO-K1 GPR34 β-Arrestin Cell Line measures GPR34 (GPCR) activity via recruitment of β-Arrestin2. This cell line ships as two vials of cryopreserved cells.

The PathHunter™ GPR34 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. lysophosphatidylserine) based activation of GPR34 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged GPR34 and EA-tagged β-Arrestin 2. Activation of GPR34 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifications

Assay Entry β-Arrestin Recruitment
Cell Line CHO-K1
For Use With (Application) Cell Lines
Primary Agonist Lyso PS
Quantity 2 Vials
Target Entry GPR34
Accession Number NM_005300.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.