missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ CHO-K1 cyno MRGPRX2 β-Arrestin Cell Line
Description
The PathHunter™ cMRGPRX2 β-Arrestin cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. PAMP-20) based activation of cMRGPRX2 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cell line overexpresses PK-tagged cMRGPRX2 and EA-tagged β-Arrestin 2. Activation of cMRGPRX2 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Cyno version of the protein.
Specifications
Specifications
| Assay Entry | β-Arrestin Recruitment |
| Cell Line | CHO-K1 |
| For Use With (Application) | Cell Lines |
| Primary Agonist | Cortistatin 14, Human |
| Quantity | 2 Vials |
| Target Entry | cMRGPRX2 |
| Accession Number | XM_015435307.1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?