Learn More
KARTELL Pasteur Pipette
Product Code. 10274991
Click to view available options
Volume (Metric):
3.5 mL
4 mL
4.4 mL
5 mL
5.4 mL
7 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 500
Drops/mL:
21
23
28
Product Code. 10274991
Brand: KARTELL 0032900
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
These disposable Pasteur Pipettes are ideal for the precise transfer of samples.
  • Disposable PE pipette with bulb and graduations.
  • The slender tip guarantees precision transfer of the sample.
  • Sterile pipettes individually wrapped.
Specifications
Material Polyethylene
Sterility Sterile
Graduated No
Color Clear
Volume (Metric) 4.4 mL
Length (Metric) 150 mm
Type Pasteur Pipet
Disposable Single-use
Fit for Purpose Sterile