Learn More
Memmert Pass-through Oven UF260TS TwinDISPLAY, 256L, 20-250°C, Forced Convection
Brand:  Memmert UF260TS
Pass-through Oven UF260TS TwinDISPLAY, 256L, 20-250°C, Forced Convection
Product Code. 16398027
  • ONLINE EXCLUSIVE PRICE
    11035.00€
    10699.00€ / Each
    Save 336.00€
    3% Off
Only null left
Estimated Shipment: 06-02-2026