Learn More
Emma Safety Footwear Paris Safety Shoes
Product Code. 15958401
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15958401
Brand: Emma Safety Footwear MM40464739
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Lightweight woven, polyurethane-coated textile with Nano Carbon Composite protective toe. Performs excellently at workplaces where safety and long-lasting comfort are top priorities. The extremely comfortable PU insole guarantees long-lasting support and cushioning of the foot.
  • S1P-Safety shoe with sole plate for dry work
  • Woven Textile
  • Polyurethane coated
  • Hydro-Tec™ Antibacterial
  • Metal-free
  • Chrome-free lining
  • ESD certified
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black, Gray
Design Paris
Liner Material Hydro-Tec™ Antibacterial
Material Textile
Size 39
Toe Material Composite