Learn More
R&D Systems™ Parameter Immunoassay Control Set 752 for TBARS
Product Code. 30001854
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001854 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30001854 Supplier R&D Systems™ Supplier No. QC164
  • ONLINE EXCLUSIVE PRICE
    210.00€
    198.45€ / Pack of 1
    Save 11.55€
    5% Off
 Restricted Item
Estimated Shipment: 11-11-2026
For use with catalog number KGE013
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Thiobarbituric Acid Reactive Substances (TBARS)
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Multi-species