missing translation for 'onlineSavingsMsg'
Learn More

Benzyl mercaptan, 99%

Product Code. 11400581
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11400581

Brand: Thermo Scientific Alfa Aesar B20081.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-53-8 | C7H8S | 124.20 g/mol

A common laboratory alkylthiol, which can be used as a source of thiol function. It also finds it uses as an odorant, synthetic flavor, and as a bacteriostat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A common laboratory alkylthiol, which can be used as a source of thiol function. It also finds it uses as an odorant, synthetic flavor, and as a bacteriostat.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-53-8
Molecular Formula C7H8S
Molecular Weight (g/mol) 124.20
MDL Number MFCD00004867,MFCD00801588,MFCD01863867
InChI Key UENWRTRMUIOCKN-UHFFFAOYSA-N
Synonym benzyl mercaptan, benzenemethanethiol, benzylthiol, alpha-toluenethiol, thiobenzyl alcohol, benzyl hydrosulfide, alpha-toluolthiol, benzylhydrosulfide, phenylmethyl mercaptan, benzylmercaptan
PubChem CID 7509
IUPAC Name phenylmethanethiol
SMILES SCC1=CC=CC=C1

Specifications

Melting Point -30°C
Density 1.053
Boiling Point 194°C to 195°C
Flash Point 70°C (158°F)
Assay Percent Range 0.99
Odor Stench
Refractive Index 1.576
Quantity 500 g
UN Number UN2810
Beilstein 605864
Sensitivity Air Sensitive
Merck Index 14,9322
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 124.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Benzyl mercaptan
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.