Learn More
Brady™ Paper Labels for M6/M7 Printers
Product Code. 17811579
Change view
Click to view available options
Dimensions (L x W):
0.9 x 0.75 in.
1 x 2 in.
1 x 3 in.
1.9 x 3 in.
1.9 x 4 in.
2 x 1 in.
For Use With (Equipment):
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
M610, M710, M611, BMP61, BMP71
M710, BMP71
Height (Metric):
22.86 mm
25.4 mm
48.26 mm
50.8 mm
Unit Size:
Pack of 100
Pack of 1000
Pack of 250
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) For Use With (Equipment) Height (Metric) unitSize
17811579 0.9 x 0.75 in. M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) 22.86 mm Pack of 250
17858114 1 x 2 in. M610, M710, M611, BMP61, BMP71 25.4 mm Pack of 1000
17808124 1 x 3 in. M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) 25.4 mm Pack of 100
17838214 1.9 x 3 in. M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) 48.26 mm Pack of 100
17821629 1.9 x 4 in. M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) 48.26 mm Pack of 100
17818854 2 x 1 in. M710, BMP71 50.8 mm Pack of 100
17848394 2 x 1 in. M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) 50.8 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17811579 Supplier Brady™ Supplier No. 173906
  • ONLINE EXCLUSIVE PRICE
    37.90€
    36.13€ / Pack of 250
    Save 1.77€
    4% Off
View all promotions
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Paper Labels with Permanent Rubber Adhesive provide good contrast and smear resistance for thermal transfer and barcode printing applications.
  • White matte paper label stock features permanent rubber adhesive
  • Low cost is ideal for all general purpose labeling projects
  • Service temperature of -40°C to 50°C (-40°F to 122°F)
  • Made with B-424 permanent adhesive
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance ASTM D 1000
Adhesive Type Permanent Rubber
Coating None
Color White
Finish Matte
Format Die-cut
Material Paper
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Barcode Labeling, Electrical Labeling, General Identification, Laboratory Labeling
For Use With (Equipment) M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
Latex Free No
Quantity 250 Labels
Shape Rectangle
Temperature Range (English) -40°F to +122°F
Temperature Range (Metric) -40°C to +50°C
Thickness (English) 0.0035 in.
Thickness (Metric) 0.089 mm
Dimensions (L x W) 0.9 x 0.75 in.
Length (English) 0.9 in.
Width (English) 0.75 in.
Width (Metric) 19.05 mm
Height (Metric) 22.86 mm
Alternate Part Number M6-80-424