Learn More
STEMCELL Technologies PancreaCult™ Organoid Growth Medium (Mouse)
Product Code. 17139231
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17139231 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17139231 Supplier STEMCELL Technologies Supplier No. 06040
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Cell culture medium for establishment and maintenance of mouse pancreatic exocrine organoids
  • PancreaCult™ Organoid Growth Medium (Mouse) is a serum-free, defined cell culture medium for establishment and maintenance of mouse pancreatic exocrine organoids
  • These organoids, or 'mini-pancreata', provide an in vitro organotypic culture system for studying pancreatic cell biology, disease, and cancer
  • Organoids grown in PancreaCult™ feature an epithelium expressing genes marking pancreatic stem cells (LGR5), progenitor cells (PDX1, SOX9), and ductal cells (CAR2, MUC1, KRT19)
  • Pancreatic organoids can be passaged every 3 - 6 days for long-term maintenance and can also be cryopreserved
  • PancreaCult™ supports mouse pancreatic organoid culture either embedded in Corning™ Matrigel™ domes or in a dilute Matrigel™ suspension
  • Organoid culture enables convenient in vitro characterization of the pancreatic epithelium in a physiologically relevant system and reduces the need for animal use
Specifications
Cell Type Pancreatic Cells
For Use With (Application) Cell Culture; Expansion; Maintenance; Organoid Culture
Quantity 1 Kit