missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Pam3CSK4, TLR1 and TLR2 Ligand

Applications: Functional, In vitro assay, In vivo assay, Ligand Activation

Brand:  Novus Biologicals™ NBP2-25297-0.025mg

Encompass

Product Code. 15779405

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »

This item is not returnable. View return policy

Description

Description

Pam3CSK4, TLR1 and TLR2 Ligand is a synthetic tripalmitoylated lipohexapeptide analog of the immunologically active N-terminal portion of bacterial lipoprotein. It activates monocytes and macrophages and is also a potent activator of proinflammatory transcription factor NF-kB. Pam3CSK4, TLR1 and TLR2 Ligand is recognized by a heterodimer formed between TLR1 and TLR2. Pam3CSK4, TLR1 and TLR2 Ligand stimulation: If your cell line does not naturally express TLR1+ TLR2, co-transfect with plasmids pCMV/TLR1and pCMV/TLR2. Forty-eight hours after transfection, stimulate cells with 10 to 100ng/ml of Pam3CSK4 for 6 to 24 hours. Determine Pam3CSK4 stimulation using appropriate detection assays. Buffer: Sterile water
Specifications

Specifications

0.5 mg/mL
0.025 mg
Human/Mouse
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Activation of TLR1 and TLR2 heterodimer. Stimulation of TLR1 and TLR2 has been reported with 10 to 100ng/mL. Use in vitro assay reported in scientific literature (PMID: 24105263) Use in functional reported in scientific literature (PMID 25511699). Use in In-vivo reported in scientific literature (PMID 25466255). Use in ligand activation reported in multiple pieces scientific literature.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.