Learn More
Paliperidone Laurate, TRC
Specifications
Specifications
| Chemical Name or Material | Paliperidone Laurate |
| CAS | 2987406-52-8 |
| Molecular Formula | C35H49FN4O4 |
| Quantity | 25 mg |
| Synonym | 3-(2-(4-(6-Fluorobenzo[d]isoxazol-3-yl)piperidin-1-yl)ethyl)-2-methyl-4-oxo-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-9-yl Dodecanoate |
| InChI Formula | InChI=1S/C35H49FN4O4/c1-3-4-5-6-7-8-9-10-11-14-32(41)43-30-13-12-20-40-34(30)37-25(2)28(35(40)42)19-23-39-21-17-26(18-22-39)33-29-16-15-27(36)24-31(29)44-38-33/h15-16,24,26,30H,3-14,17-23H2,1-2H3 |
| SMILES | O=C1N2CCCC(OC(CCCCCCCCCCC)=O)C2=NC(C)=C1CCN3CCC(C4=NOC5=C4C=CC(F)=C5)CC3 |
| IUPAC Name | 3-(2-(4-(6-fluorobenzo[d]isoxazol-3-yl)piperidin-1-yl)ethyl)-2-methyl-4-oxo-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-9-yl dodecanoate |
| Molecular Weight (g/mol) | 608.79 |
| Formula Weight | 608.37 |