Learn More
Paliperidone-d4 (Major), TRC
Product Code. 30421804
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421804 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30421804 Supplier Toronto Research Chemicals Supplier No. TRCP14100310MG
 Restricted Item
Estimated Shipment: 14-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material rac 9-Hydroxy Risperidone-d4
CAS 1020719-55-4
Molecular Formula C23 2H4 H23 F N4 O3
Quantity 10 mg
Synonym Paliperidone-(ethyl-D4), 9-Hydroxyrisperidone-(ethyl-D4)
InChI Formula InChI=1S/C23H27FN4O3/c1-14-17(23(30)28-9-2-3-19(29)22(28)25-14)8-12-27-10-6-15(7-11-27)21-18-5-4-16(24)13-20(18)31-26-21/h4-5,13,15,19,29H,2-3,6-12H2,1H3/i8D2,12D2
SMILES [2H]C([2H])(N1CCC(CC1)c2noc3cc(F)ccc23)C([2H])([2H])C4=C(C)N=C5C(O)CCCN5C4=O
IUPAC Name 9-hydroxy-2-methyl-3-[1,1,2,2-tetradeuterio-2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
Molecular Weight (g/mol) 430.51
Formula Weight 430.2318