Learn More
Thermo Scientific™ PageRuler™ Unstained Low Range Protein Ladder
Product Code. p-7225564 Shop All Thermo Scientific Products
Click to view available options
Quantity:
2 x 250 μL
Unit Size:
Pack of 2
Product Code. 11802134
Brand: Thermo Scientific™ 26632
Edge
Thermo Scientific PageRuler Unstained Low Range Protein Ladder is a mixture of eight purified proteins and peptides (3.4 to 100 kDa) for use as size standards in peptide and small-protein gel electrophoresis.
Thermo Scientific™ PageRuler Unstained Low Range Protein Ladder is a mixture of eight (8) purified proteins and peptides (3.4 to 100kDa) for use as size standards in peptide and small protein gel electrophoresis.
Thermo Scientific PageRuler Unstained Low Range Protein Ladder is a mixture of eight purified proteins and peptides (3.4 to 100 kDa) for use as size standards in peptide and small-protein gel electrophoresis. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.
Product features
  • Reference bands—25 kDa band is stronger than the others for easy orientation
  • Tagged—the ladder proteins (except for the 5 and 3.4 kDa peptides) contains an integral Strep-tag™ II sequence and can be detected on western blots using Strep-Tactin™ conjugates
Applications
  • Accurate protein sizing on SDS-polyacrylamide gels and western blots
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Includes two vials of 250 μL each
Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 100 mM DTT, 1 mM NaN3, 0.01% bromophenol blue and 33% glycerol
Upon receipt, store at -20°C.
Number of Markers 8
Product Type Protein Ladder
Ready to Load Yes
Recommended Applications Tricine gels
Size Range 3.4 to 100 kDa
Stain Type Unstained
For Use With (Application) Tricine gels
Gel Compatibility Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Molecular Weight (g/mol) 100, 30, 25, 20, 15, 10, 5, 3.4 kDa
Product Line PageRuler
Quantity 2 x 250 μL
Shipping Condition Approved for shipment on Wet or Dry Ice
System Type SDS-PAGE