missing translation for 'onlineSavingsMsg'
Learn More

Paclitaxel 7-((alphaR,betaS)-beta-(Benzoylamino)-alpha-hydroxybenzenepropanoate, TRC

Product Code. 30332704
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30332704 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332704 Supplier Toronto Research Chemicals Supplier No. TRCB00200510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Paclitaxel 7-(alphaR,betaS)-beta-(Benzoylamino)-alpha-hydroxybenzenepropanoate
CAS 376629-82-2
Molecular Formula C63 H64 N2 O17
Quantity 10 mg
Synonym (2aR,4S,4aS,6R,9S,11S,12S,12aR,12bS)-6,12b-Bis(acetyloxy)-12-(benzoyloxy)-2a,3,4,4a,5,6,9,10,11,12,12a,12b-dodecahydro-11-hydroxy-4a,8,13,13-tetramethyl-5-oxo-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxete-4,9-diylester-(αR,α'R,βS,β'S)-β-(benzoylamino)-α-hydroxy-benzenepropanoic Acid
InChI Formula InChI=1S/C63H64N2O17/c1-35-43(79-58(74)49(68)47(38-22-12-7-13-23-38)64-55(71)40-26-16-9-17-27-40)33-63(76)54(81-57(73)42-30-20-11-21-31-42)52-61(6,53(70)51(78-36(2)66)46(35)60(63,4)5)44(32-45-62(52,34-77-45)82-37(3)67)80-59(75)50(69)48(39-24-14-8-15-25-39)65-56(72)41-28-18-10-19-29-41/h7-31,43-45,47-52,54,68-69,76H,32-34H2,1-6H3,(H,64,71)(H,65,72)/t43-,44-,45+,47-,48-,49+,50+,51+,52-,54-,61+,62-,63+/m0/s1
SMILES CC(=O)O[C@H]1C(=O)[C@]2(C)[C@H](C[C@H]3OC[C@@]3(OC(=O)C)[C@H]2[C@H](OC(=O)c4ccccc4)[C@]5(O)C[C@H](OC(=O)[C@H](O)[C@@H](NC(=O)c6ccccc6)c7ccccc7)C(=C1C5(C)C)C)OC(=O)[C@H](O)[C@@H](NC(=O)c8ccccc8)c9ccccc9
IUPAC Name [(1S,2S,3R,4S,7R,9S,10S,12R,15S)-4,12-diacetyloxy-9,15-bis[[(2R,3S)-3-benzamido-2-hydroxy-3-phenylpropanoyl]oxy]-1-hydroxy-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.03,10.04,7]heptadec-13-en-2-yl] benzoate
Molecular Weight (g/mol) 1121.186
Formula Weight 1120.42
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.