Learn More
Thermo Scientific™ µPAC™ Neo HPLC Columns
Product Code. 17887883 Shop All Thermo Scientific Products
Change view
Click to view available options
Unit Size:
Each
Includes:
110 cm μPAC™ Neo HPLC Column
110 cm µPAC Neo Plus column
50 cm μPAC™ Neo HPLC Column
50 cm µPAC Neo Plus column
HT µPAC Neo Plus column
Product Line:
µPAC Neo
µPAC Neo Plus
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize Includes Product Line
17887883 Each - -
17817893 Each 50 cm μPAC™ Neo HPLC Column µPAC Neo
17897883 Each 110 cm μPAC™ Neo HPLC Column µPAC Neo
30804779 Each HT µPAC Neo Plus column µPAC Neo Plus
30804774 Each 50 cm µPAC Neo Plus column µPAC Neo Plus
30804780 Each 110 cm µPAC Neo Plus column µPAC Neo Plus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17887883 Supplier Thermo Scientific™ Supplier No. COLLOLO050NEOB
Learn more about your proteome samples with Thermo Scientific™ μPAC™ Neo Plus HPLC columns.
Learn more about your proteomics samples with Thermo Scientific™ μPAC™ Neo Plus HPLC columns. Taking your performance even further, our μPAC Neo Plus column line offers higher resolution than the previous generation of μPAC™ Neo columns, ensuring that you can extract the maximum amount of information from highly complex samples using low-flow chromatography. Column installation is simple and reliable with Thermo Scientific™ nanoViper™ Fingertight Fittings.
The μPAC Neo Plus column range is the ideal choice for a variety of proteomics experiments, including fast label-free quantitation, comprehensive identification, and single cell analysis.
Performance and Robustness - μPAC Neo columns operate at moderate pressures, allowing for longer flow paths that increase separation performance and sensitivity. Operating at lower back pressures also extends column lifetime.
Column-to-Column Reproducibility - The unique micromachining manufacturing of our μPAC Neo column line produces columns that are virtually identical. This results in increased injection-to-injection and column-to-column reproducibility, providing you with greater confidence in your results throughout the duration of a research study.
Perfect Flow Path Order - μPAC Neo columns are designed with a perfect flow path order, eliminating heterogeneous flow paths. The ordered flow path of the columns minimizes dispersion to the overall separation, resulting in sharper and more intense chromatographic peaks.
TRUSTED_SUSTAINABILITY
Specifications
Flow Rate 0.1 to 0.75 μL/min
Max. Pressure 6526 psi (450 bar)
pH 1.5 to 7
Pore Size 100 to 300 Å
Porosity 59%
Temperature 60°C
Stationary Phase C18
Packing Material Silicon Chip
Column Type Cylindrical Micro Pillar Array
Endcapped Yes
Phase Reversed Phase
Description 50 cm Low-Load μPAC™ Neo HPLC Column
Bed Length 50 cm
Bed Width 180 μm
Interpillar Distance 1.25 μm
Pillar Diameter 2.5 μm
Pillar Length 30 μm