Learn More
Thermo Scientific™ μPAC™ Neo Plus ESI-MS Accessories
Product Code. 17827893 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Grounding Cable
Interface
Voltage Spacer
Length (Metric):
500 mm
60 mm
N/A
Diameter (Metric) Inner:
10 μm
20 μm
N/A
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Length (Metric) Diameter (Metric) Inner unitSize
17827893 Interface N/A N/A Each
30804772 Voltage Spacer 60 mm 10 μm Each
30804775 Voltage Spacer 60 mm 20 μm Each
30804776 Voltage Spacer 500 mm 20 μm Each
30804777 Interface 60 mm 10 μm Each
30804773 Interface 60 mm 20 μm Each
30804778 Grounding Cable N/A N/A Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17827893 Supplier Thermo Scientific™ Supplier No. EMIFLEXIONB
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
Use μPAC™ ESI-MS Accessories for fast, reproducible, and low dispersion grounded connection between Thermo Scientific™ μPAC™ Neo Plus HPLC columns and Thermo Scientific™ Nanospray Flex™ and EASY-Spray™ ion sources.
Use Thermo Scientific™ μPAC™ ESI-MS Accessories to facilitate reproducible, low dispersion, and grounded connections between μPAC Neo Plus HPLC columns and low-flow mass spectrometer ion sources. μPAC Neo Plus voltage spacers are available for both EASY-Spray and Nanospray Flex ion sources. All configurations require a grounding cable.
The μPAC Flex iON Connect ESI-MS Interface consists of two separate units:
  • A docking unit that can be slid onto the Nanospray Flex Ion Source's metal rod and receives both a grounding and a high voltage cable
  • A spray unit compatible with the outlet fittings of μPAC columns on one side and compatible with any 360 μm OD emitter on the other side.
The μPAC EASY-Spray emitter is designed for easy connection to an EASY-Spray MS Source. It contains a pre-assembled EASY-Spray emitter with a transfer line and grounding union.
TRUSTED_SUSTAINABILITY
Specifications
Description μPAC™ Flex iON Connect ESI-MS Interface
For Use With (Equipment) Nanospray Flex™ Ion Source
Type Interface
Length (Metric) N/A
Diameter (Metric) Inner N/A