missing translation for 'onlineSavingsMsg'
Learn More

p65/RELA Mouse anti-Human, Rat, Clone: 1B12D11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66535-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16818505

  • 399.00€ / 150µL
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

This gene encodes subunit 1 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 1 belongs to the SURP protein family; named for the SURP motifs that are thought to mediate RNA binding. Subunit 1 has tandemly repeated SURP motifs in its amino-terminal half while its carboxy-terminal half contains a proline-rich region and a ubiquitin-like domain. Binding studies with truncated subunit 1 derivatives demonstrated that the two SURP motifs are necessary for binding to subunit 3 while contacts with subunit 2 may occur through sequences carboxy-terminal to the SURP motifs. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications

Specifications

p65/RELA
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q04206
Rela
p65; RELA Fusion Protein Ag1199
150 μL
Primary
Human, Rat
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1B12D11
Unconjugated
RELA
NFKB3, p65, RELA, Transcription factor p65
Mouse
Protein G
RUO
309165, 5970
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers