missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2Y11/P2RY11 Mouse anti-Human, Alexa Fluor 594, Clone: 505214, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9305T-100UG
This item is not returnable.
View return policy
Description
P2Y11/P2RY11 Monoclonal specifically detects P2Y11/P2RY11 in Human samples. It is validated for Flow Cytometry.Specifications
| P2Y11/P2RY11 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| P2RY11, P2Y11P2Y purinoceptor 11, purinergic receptor P2Y, G-protein coupled, 11, purinergic receptor P2Y11 | |
| NS0 mouse myeloma cell line transfected with human P2Y11/P2RY11 Accession # Q96G91 | |
| RUO | |
| 5032 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 505214 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q96G91 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Stains human P2Y11/P2RY11 transfectants but not irrelevant transfectants in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only (RUO)