Learn More
Melford P-Nitrophenyl-B-D-Fucopyranoside
Product Code. 30018630
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018630 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30018630 Supplier Melford Supplier No. N820401.0
 Restricted Item
Estimated Shipment: 28-10-2026
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Free PNP: <100ppm
Specifications
Quantity 1 g