Learn More
p-Nitrophenyl Alpha-L-Fucopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Nitrophenyl-alpha-L-fucopyranoside |
| CAS | 10231-84-2 |
| Molecular Formula | C12 H15 N O7 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C12H15NO7/c1-6-9(14)10(15)11(16)12(19-6)20-8-4-2-7(3-5-8)13(17)18/h2-6,9-12,14-16H,1H3/t6-,9+,10+,11-,12-/m0/s1 |
| SMILES | C[C@@H]1O[C@@H](Oc2ccc(cc2)[N+](=O)[O-])[C@@H](O)[C@H](O)[C@@H]1O |
| IUPAC Name | (2S,3S,4R,5S,6S)-2-methyl-6-(4-nitrophenoxy)oxane-3,4,5-triol |
| Molecular Weight (g/mol) | 285.25 |
| Formula Weight | 285.0849 |