missing translation for 'onlineSavingsMsg'
Learn More

p-Coumaryl Alcohol, TRC

Product Code. 30366674
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366674 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30366674 Supplier Toronto Research Chemicals Supplier No. TRCC755420500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material p-Coumaryl Alcohol
CAS 3690-05-9
Purity Grade Notes HPLC
Molecular Formula C9 H10 O2
Quantity 500 mg
Synonym Phenol, 4-(3-hydroxy-1-propen-1-yl)-, 2-Propen-1-ol, 3-(p-hydroxyphenyl)- (6CI,7CI,8CI), Phenol, 4-(3-hydroxy-1-propenyl)- (9CI), 3-(p-Hydroxyphenyl)-2-propen-1-ol, 4-(3-Hydroxy-1-propen-1-yl)phenol, 4-Hydroxycinnamic alcohol, 4-Hydroxycinnamyl alcohol, p-Coumaric alcohol, p-Coumaryl alcohol, p-Cumaric alcohol, p-Hydroxycinnamic alcohol, p-Hydroxycinnamyl alcohol
InChI Formula InChI=1S/C9H10O2/c10-7-1-2-8-3-5-9(11)6-4-8/h1-6,10-11H,7H2/b2-1+
SMILES OC\C=C\c1ccc(O)cc1
IUPAC Name 4-[(E)-3-hydroxyprop-1-enyl]phenol
Molecular Weight (g/mol) 150.17
Formula Weight 150.0681
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.